6-Carboxy-2',4,4',5',7,7'-hexachlorofluorescein succinimidyl ester (6-HEX SE) single isomer ≥90%
Catalog # AATB202
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:680.06 g/mol
CAS Number:2129651-79-0
Formula:C₂₅H₉Cl₆NO₉