Order Entry
Italy
ContactUsLinkComponent
ICG acid ≥95%
ICG acid ≥95%
Catalog # AATB189
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
ICG acid ≥95%
Catalog # AATB189
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :844.98 g/mol