Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Propidium Iodide ≥95%

Irritant
Supplier:   AAT Bioquest
Propidium Iodide ≥95%
Supplier:  AAT Bioquest
Certificates
Propidium Iodide ≥95%
Propidium Iodide ≥95%

2 Options Available Below

Product Details & Documents

Propidium iodide (PI) belongs to the same chemical class of ethidium bromide.

Synonyms:  3,8-Diamino-5-[3-(diethylmethylammonio)propyl]-6-phenylphenanthridinium diiodide, PI, 3,8-Diamino-5-(3-(diethyl(methyl)ammonio)propyl)-6-phenylphenanthridin-5-ium iodide, 3,8-Diamino-5-(3-diethylaminopropyl)-6-phenylphenanthridinium iodide methiodide
  • CAS Number:25535-16-4
  • Formula:C₂₇H₃₄I₂N₄
  • MDL Number:MFCD00011921
  • Molecular Weight:668.4 g/mol
  • ADR:5,II
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 156,00 €
    156,00 €
    1 * 25 mg
       156,00 €

  • Plastic bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 6 240,00 €
    6 240,00 €
    1 * 5 g
       6 240,00 €