Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
TF1-dUTP
TF1-dUTP 1 mM in TRIS buffer (pH 7.5)
TF1-dUTP 1 mM in TRIS buffer (pH 7.5)
Irritant
TF1-dUTP 1 mM in TRIS buffer (pH 7.5)
Catalog # AATB17006
Supplier:
AAT Bioquest
CAS Number:
Expand image
Irritant
TF1-dUTP 1 mM in TRIS buffer (pH 7.5)
Catalog # AATB17006
Supplier:
AAT Bioquest
CAS Number:
Specifications
Pack type:
Plastic bottle
Pk:
25 Nmo
Seq:
0311
Specifications
About this item
Product Details
:
:
1000 g/mol
ADR:
5,II
:
GHS07
Product Details