Order Entry
Italy
ContactUsLinkComponent
Analogue stopwatches, Amigo
Analogue stopwatches, Amigo
Catalog # 609-1240
Supplier:  HANHART
undefined
Analogue stopwatches, Amigo
Catalog # 609-1240
Supplier:  HANHART
Specifications
  • Ø:
    55 mm
  • Colour:
    Black
  • Description:
    Timer, 1/10 s, 15 min.
  • Pk:
    1
Specifications
About this item
Analogue stopwatches with robust ABS casing.
  • Pin lever, one jewel
  • ¹/₁₀ second
  • 15 minutes capacity
  • Shock-protected pin lever movement
  • Robust ABS case, with cord for hanging round the neck