Sieves for Pilotina Mills
Supplier: IKA
Certificates
About this item
Interchangeable sieves for insertion into the grinding heads ensure maximum particle size filtering.
All sieves are made of stainless steel 1.4301.
Frequently Bought Together
Interchangeable sieves for insertion into the grinding heads ensure maximum particle size filtering.
All sieves are made of stainless steel 1.4301.