Order Entry
Italy
ContactUsLinkComponent
5,7-Dihydroxy-2-(4-hydroxyphenyl)-6-((2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)-4H-chromen-4-one, yellow, solid
5,7-Dihydroxy-2-(4-hydroxyphenyl)-6-((2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)-4H-chromen-4-one, yellow, solid
Catalog # EHERC14479800-10MG
Supplier:  EHRENSTORFER
CAS Number:  
5,7-Dihydroxy-2-(4-hydroxyphenyl)-6-((2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)-4H-chromen-4-one, yellow, solid
Catalog # EHERC14479800-10MG
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :432.38 g/mol
Melting Pt:235...239 °C
CAS Number:38953-85-4
Formula:C₂₁H₂₀O₁₀
MDL Number:MFCD00076044