Conjugation kits, protein-oligo conjugation kit
VECTS-9011-1
:
- Pk:1 KIT
- Description:Protein-Oligo Conjugation kit (2 conjugations)
- Environmentally Preferable:
The Protein-Oligo Conjugation kit is designed to easily and efficiently conjugate two separate protein-oligo conjugates. This kit is flexible so that researchers with little or no conjugation experience can make their own custom protein-oligo conjugates to suit their needs. It includes all of the necessary components - including S-HyNic and S-4FB - and protocols for easy and specific crosslinking of any protein with any oligo up to 100 base pairs in length.
- Catalysed conjugation – Faster kinetics for greater efficiency and yields
- Quantifiable – Using a UV signature wavelength and simple UV scan
- Stable – 10 times more stable than any other conjugation linker
- Specificity – Two-linker method avoids homoconjugate formation