Order Entry
Italy
ContactUsLinkComponent
Measles Virus IgM (Human) ELISA kit
Measles Virus IgM (Human) ELISA kit
Catalog # ABNOKA6782.1KIT
Supplier:  Abnova
CAS Number:  
Measles Virus IgM (Human) ELISA kit
Catalog # ABNOKA6782.1KIT
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Primary antibody reactivity:
    Human
  • Target protein:
    IgM
  • Assay Principle:
    Qualitative

Specifications

About this item

Measles Virus IgM (Human) ELISA Kit is a qualitative determination of IgM class antibodies against Measles Virus in human serum or plasm.