Order Entry
Italy
ContactUsLinkComponent
Mumps Virus IgM (Human) ELISA kit
Mumps Virus IgM (Human) ELISA kit
Catalog # ABNOKA6784.1KIT
Supplier:  Abnova
CAS Number:  
Mumps Virus IgM (Human) ELISA kit
Catalog # ABNOKA6784.1KIT
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Primary antibody reactivity:
    Human
  • Target protein:
    IgM
  • Assay Principle:
    Qualitative

Specifications

About this item

Mumps Virus IgM (Human) ELISA Kit is a qualitative determination of IgM class antibodies against Mumps Virus in human serum or plasma.