Cytomegalovirus IgM ELISA kit
Catalog # ABNOKA6707.1KIT
Supplier: Abnova
CAS Number:
Specifications
- Pk:1 KIT
- Primary antibody reactivity:Cytomegalovirus
- Target protein:IgM
- Assay Principle:Quantitative
Specifications
About this item
Cytomegalovirus IgM ELISA Kit is an enzyme immunoassay for the quantitative and qualitative measuremnet of IgM antibodies to Cytomegalovirus (CMV) in human serum and plasma.