Order Entry
Italy
ContactUsLinkComponent
Synthetic Yeast Inorganic Pyrophosphatase (from E. coli)
Synthetic Yeast Inorganic Pyrophosphatase (from E. coli)
Catalog # ABNOP9370.100UL
Supplier:  Abnova
CAS Number:  
Synthetic Yeast Inorganic Pyrophosphatase (from E. coli)
Catalog # ABNOP9370.100UL
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    100 µl
  • Enzyme type:
    Synthetic
  • Source:
    E. coli
  • Species:
    Yeast
  • Enzyme name:
    Inorganic Pyrophosphatase

Specifications

About this item