Recombinant Mouse CXCL10 (from E. coli)
25 µG
Catalog # ABNOP9491.25UG
Supplier: Abnova
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:25 µG
- Cat. No.:ABNOP9491.25UG
- Protein/peptide type:Recombinant
- Protein/peptide name:CXCL10
- Gene ID:P17515
- Sequence:22a.a.-98a.a.
- Species:Mouse
- Source:E. coli



