Recombinant Guinea pig CXCL10 (from E. coli)
Catalog # ABNOP9494.50UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Guinea pig
- Gene ID:A0A286XCT1
- Protein/peptide name:CXCL10
- Sequence:22a.a.-97a.a.