Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Recombinant Human HGF (from E. coli)

20 µG
  ABNOP8761.20UG
 :   Abnova
Recombinant Human HGF (from E. coli)
  ABNOP8761.20UG
 :  Abnova
 :  
Recombinant Human HGF (from E. coli)

20 µG

 :  
 
264,00 €
   
1 * 20 µG
 
 

 

  • Pk:
    20 µG
  • Cat. No.:
    ABNOP8761.20UG
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    HGF
  • Species:
    Human
  • Source:
    E. coli