Recombinant Human FGF8 (from E. coli)
25 µG
Catalog # ABNOP8603.25UG
Supplier: Abnova
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:25 µG
- Cat. No.:ABNOP8603.25UG
- Protein/peptide type:Recombinant
- Protein/peptide name:FGF8
- Species:Human
- Source:E. coli



