Recombinant Human FGFR2 (from CHO cells)
Catalog # ABNOP7606.50UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:CHO cells
- Species:Human
- Gene ID:P21802
- Protein/peptide name:FGFR2
- Sequence:22a.a.-289a.a.