Anti-Histone H14 Rabbit Recombinant Antibody [clone: 30]
ABNORAB04185.100UL
: Abnova
- Antigen symbol:Histone H14
- Clonality:Recombinant
- Clone:30
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human Histone H1.4
- Pk:100 µl