Order Entry
Italy
ContactUsLinkComponent
Anti-EIF5A Rabbit Recombinant Antibody [clone: 50]
Anti-EIF5A Rabbit Recombinant Antibody [clone: 50]
Catalog # ABNORAB03991.100UL
Supplier:  Abnova
Anti-EIF5A Rabbit Recombinant Antibody [clone: 50]
Catalog # ABNORAB03991.100UL
Supplier:  Abnova

Specifications

  • Antigen symbol:
    EIF5A
  • Clonality:
    Recombinant
  • Clone:
    50
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human EIF5A.
  • Pk:
    100 µl

Specifications

About this item