Order Entry
Italy
ContactUsLinkComponent
Anti-C5AR1 Rabbit Recombinant Antibody [clone: 32-G1]
Anti-C5AR1 Rabbit Recombinant Antibody [clone: 32-G1]
Catalog # ABNORAB03112.200UG
Supplier:  Abnova
Anti-C5AR1 Rabbit Recombinant Antibody [clone: 32-G1]
Catalog # ABNORAB03112.200UG
Supplier:  Abnova

Specifications

  • Antigen symbol:
    C5AR1
  • Clonality:
    Recombinant
  • Clone:
    32-G1
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human C5AR1.
  • Pk:
    200 µG

Specifications

About this item