Order Entry
Italy
ContactUsLinkComponent
Anti-ANXA1 Rabbit Recombinant Antibody [clone: SAIC-13B-19]
Anti-ANXA1 Rabbit Recombinant Antibody [clone: SAIC-13B-19]
Catalog # ABNORAB03664.200UG
Supplier:  Abnova
Anti-ANXA1 Rabbit Recombinant Antibody [clone: SAIC-13B-19]
Catalog # ABNORAB03664.200UG
Supplier:  Abnova

Specifications

  • Antigen symbol:
    ANXA1
  • Clonality:
    Recombinant
  • Clone:
    SAIC-13B-19
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human ANXA1.
  • Pk:
    200 µG

Specifications

About this item