Order Entry
Italy
ContactUsLinkComponent
Anti-Cytokeratin 18 Rabbit Recombinant Antibody [clone: R04-1A7]
Anti-Cytokeratin 18 Rabbit Recombinant Antibody [clone: R04-1A7]
Catalog # ABNORAB01680.100UL
Supplier:  Abnova
Anti-Cytokeratin 18 Rabbit Recombinant Antibody [clone: R04-1A7]
Catalog # ABNORAB01680.100UL
Supplier:  Abnova

Specifications

  • Antigen symbol:
    Cytokeratin 18
  • Clonality:
    Recombinant
  • Clone:
    R04-1A7
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against synthetic peptide of human Cytokeratin 18
  • Pk:
    100 µl

Specifications

About this item