Order Entry
Italy
ContactUsLinkComponent
Anti-Cytokeratin 16 Rabbit Recombinant Antibody [clone: R02-5F8]
Anti-Cytokeratin 16 Rabbit Recombinant Antibody [clone: R02-5F8]
Catalog # ABNORAB01704.100UL
Supplier:  Abnova
Anti-Cytokeratin 16 Rabbit Recombinant Antibody [clone: R02-5F8]
Catalog # ABNORAB01704.100UL
Supplier:  Abnova

Specifications

  • Antigen symbol:
    Cytokeratin 16
  • Clonality:
    Recombinant
  • Clone:
    R02-5F8
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human Cytokeratin 16
  • Pk:
    100 µl

Specifications

About this item