Anti-TAG-72 Mouse Recombinant Antibody [clone: CC49 (MINRETUMOMAB)]
Catalog # ABNORAB01180.200UG
Supplier: Abnova
Specifications
- Antigen symbol:TAG-72
- Clonality:Recombinant
- Clone:CC49 (MINRETUMOMAB)
- Host:Mouse
- Reactivity:Human
- Immunogen:Recombinant mouse monoclonal antibody raised against human TAG-72.
- Pk:200 µG