Order Entry
Italy
ContactUsLinkComponent
Anti-TAG-72 Rabbit Recombinant Antibody [clone: CC49 (MINRETUMOMAB)]
Anti-TAG-72 Rabbit Recombinant Antibody [clone: CC49 (MINRETUMOMAB)]
Catalog # ABNORAB01181.200UG
Supplier:  Abnova
Anti-TAG-72 Rabbit Recombinant Antibody [clone: CC49 (MINRETUMOMAB)]
Catalog # ABNORAB01181.200UG
Supplier:  Abnova

Specifications

  • Antigen symbol:
    TAG-72
  • Clonality:
    Recombinant
  • Clone:
    CC49 (MINRETUMOMAB)
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human TAG-72.
  • Pk:
    200 µG

Specifications

About this item