Protein InstaStain® for 15 gels
Catalog # EDVO2016
Supplier: EDVOTEK
CAS Number:
Specifications
- Pk:15
- Description:Protein InstaStain® for 15 gels
Specifications
About this item
Protein InstaStain® sheets stain gels faster than conventional methods.
- For 15 gels (10×10 cm)
Protein InstaStain® gives high quality and uniform gel staining with excellent results for photography. They are also environmentally friendly because they use a solid matrix, avoiding large amounts of liquid stain and waste disposal.
Protein InstaStain® can be used to stain the most common gels of various sizes (ranging from 8×8 to 13×13 cm).