COURIER™ 3000 Shipping Scales
Supplier: OHAUS
Certificates
About this item
The Courier 3000 Series shipping scales intuitive, user-friendly interfaces, bright displays, and low-profile steel construction ensure durability and reliability from offices to warehouses to even veterinary offices (the Courier 3000 is effective for weighing small animals). Versatile and effective.
- For Weighing and display hold
- 4-digit, 7-segment LCD display with white backlight, 28 mm high digits
- ABS indicator, powder-coated steel platform and frame
- 150% overload protection
- AC adapter included
- 100 Hours with disposable batteries
- Painted steel platform construction
Specifications
- Catalog No:
- OHAU30809914
- OHAU30746023
- OHAU30809915
- OHAU30746025
- OHAU30809912
- OHAU30746022
- OHAU30809913
- OHAU30809911
- OHAU30746021
- Operating temperature range:
- −10...+40 °C
- −10...+40 °C
- −10...+40 °C
- −10...+40 °C
- −10...+40 °C
- −10...+40 °C
- −10...+40 °C
- −10...+40 °C
- −10...+40 °C
- Voltage:
- 100 - 240 VAC
- 100 - 240 VAC
- 100 - 240 VAC
- 100 - 240 VAC
- 100 - 240 VAC
- 100 - 240 VAC
- 100 - 240 VAC
- 100 - 240 VAC
- 100 - 240 VAC