Order Entry
Italy
ContactUsLinkComponent
Laboratory Glassware, Basket kit, Model APLW 214
Laboratory Glassware, Basket kit, Model APLW 214
  MIEL12036820
 :  MIELE
Laboratory Glassware, Basket kit, Model APLW 214
  MIEL12036820
 :  MIELE

 

  • Description:
    Basket kit, Stainless steel
    Plastic, Black
  • W×D×H:
    560×590×450 mm
  • Weight:
    10,33 kg
  • Pk:
    1 SET

 

 

Basket kit for the treatment of laboratory glassware with injector nozzles & inserts.

  • To place 36 narrow-necked glassware and other glassware
  • It consists of: upper basket A 101 and inf A 150
  • With A 11/2 insert and 2 A 301/5 modules
  • Component of the EasyLoad system