96 Well Plate, RCH CEREX PSAX
Plate RCH Cerex PSAX; 1 ml, 96, PSAX , 10 mg
Catalog # TCAT615-0101RCH
Supplier: Tecan
Jump to:
Product Details & Documents
A 17 µm polymer micro particulate with strong anionic exchange chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and uniform sample prep.
- Cerex
- Ion-exchange
- Liquid handler compatible
Ideal for acidic target compounds.
Packaging: 1 piece per box
Specifications
- No. of wells:96
- Description:Plate RCH Cerex PSAX; 1 ml
- Phase:PSAX
- Filling:10 mg
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



