Sterile Sleeve/Glove System
PIECM200/90EBE4
: PIERCAN
- Anti-static:+
- Material:EPDM rubber
- Powdered / powder-free:Powder-free
- Colour:Black
- Description:Sleeve/glove system
- Length:550 mm
- Tensile strength:>18 MPa
- Pk:10
Black EPDM (Ethylene Propylene Diene Monomer) sleeve. Excellent chemical properties, good mechanical properties. Composition in accordance with the FDA positive list (CFR 21 § 177.2600).
- Standard powder-free sleeve
- Non-recyclable
- Temperature range: –30 to +120 °C
Store in the original packaging between 5 and 25 °C. Expiry date of 3 years in the original packaging. Wash with soapy water, rinse with clear water and then air-dry at a low temperature (<40 °C). Excellent resistance to the following disinfectants: 0,25% Surfanios, 0,50% Spetanios, 10% Soproper, 100% Soproper, 70% Isopropyl alcohol. Disposal as simple non-toxic waste if not contaminated by a hazardous substance.
Thickness: 4/10 mm (palm min: 3/10, max: 6/10), Length tolerance (±20 mm), Diameter of the rim: 5 mm (±1 mm).
: Individually packed in two-toned black and transparent, heat-sealed and easy-opening pouch. Pack of 10 units.