Foam insert for 3M™ Solus™ 1000 spectacles
Foam insert for 3M™ Solus™ 1000 spectacles, for a closer fit to the face
Catalog # MMMASOLUSSR
Supplier: 3M
Jump to:
Product Details & Documents
Foam insert designed for use with 3M™ Solus™ 1000 safety spectacles.
- Fills the gap between the eyewear and the wearer's face, further reducing the risk of injury from projectiles and other debris
- Offers extra comfort and protection from sweat, ideal for hot and humid conditions
Specifications
- Description:Foam insert for 3M™ Solus™ 1000 spectacles, for a closer fit to the face