Heat Resistant Polyimide Labels for M710 and BMP71
: Brady
High Temperature Polyimide (B-727) withstands wave solder environments and multiple harsh washes for printed circuit board and electronic component preprocess labelling.
- Ultra-durable adhesive is designed for use with extreme wash protocol and cleaning chemicals
- Ideal for use in auto apply equipment
- Glossy white polyimide material is thermal transfer printable
- Service temperature of −70 to 100 °C (−94 to 212 °F)
Ideal for circuit board labelling, component and equipment labelling, and temperature indicating labelling.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M7-R6000.
: UL recognised. Meets the performance requirements of MIL -STD-202G, Notice 12, Method 215K. Dibutyltin- and Dioctyltin-free.
Frequently Bought Together