Order Entry
Italy
ContactUsLinkComponent
Organic Reference Standard, 8-Hydroxyquinoline sulphate, Dr Ehrenstorfer
Organic Reference Standard, 8-Hydroxyquinoline sulphate, Dr Ehrenstorfer
Catalog # EHERC14250000
Supplier:  EHRENSTORFER
CAS Number:  
Organic Reference Standard, 8-Hydroxyquinoline sulphate, Dr Ehrenstorfer
Catalog # EHERC14250000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pk:
    0,25 g
  • Standards:
    Reference standard
  • CAS No.:
    134-31-6
  • Description:
    8-Hydroxyquinoline sulphate
  • Formula:
    2C₉H₇NO.H₂O₄S
  • Molecular weight:
    388.39 g/mol
  • Description:
    8-Hydroxyquinoline sulfate

Specifications

About this item

Certifications: Meets ISO/IEC 17025, ISO 17034