Culti-Loops™ Bacteroides fragilis ATCC® 25285™
Catalog # OXOIR4601250
Supplier: THERMO FISHER DIAGNOSTICS
CAS Number:
Specifications
- Pk:5
- Description:Bacteroides fragilis ATCC® 25285™
- Seq:53
Specifications
About this item
Culti-Loops™ QC organisms. Bacteroides fragilis ATCC® 25285™ is recommended to be used for culture media and antimicrobial susceptibility quality control testing.
Bacteroides fragilis ATCC® 25285™ is recommended to be used for culture media and antimicrobial susceptibility quality control testing by CLSI. β-lactamase positive strain. The loop may be dissolved in liquid medium or streaked directly onto an appropriate media.
Packaging: Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.