Order Entry
Italy
ContactUsLinkComponent
Poloxamer 181 (Pluronic® L-61, Synperonic® L61) surfactant
Poloxamer 181 (Pluronic® L-61, Synperonic® L61) surfactant
Catalog # SPCMP1162-500MLGL
Supplier:  Spectrum Chemical
CAS Number:  
Poloxamer 181 (Pluronic® L-61, Synperonic® L61) surfactant
Catalog # SPCMP1162-500MLGL
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Cloud Point (10 % Aqueous):
    15 - 19 °C
  • pH of a 2.5% Aqueous Solution:
    5.0 - 7.5
  • Color (APHA):
    max. 50
  • Water:
    max. 0.4 %

Specification Test Results

About this item

 :
 :2000 g/mol
Storage Temperature:Ambient
CAS Number:9003-11-6
Formula:HO(C₂H₄O)a(C₃H₆O)b(C₂H₄O)a
MDL Number:MFCD00082049