Order Entry
Italy
Orders LinkContactUsLinkComponent
Acid red 52

Acid red 52

5 kg
Catalog # SPCMA1586-5KGBL
Supplier:   Spectrum Chemical
Acid red 52
Catalog # SPCMA1586-5KGBL
Supplier:  Spectrum Chemical
CAS Number:  3520-42-1
Acid red 52

5 kg

CAS Number:  3520-42-1
Price
2 810,00 €
Log in for availability
1 * 5 kg
Product Details & Documents

Acid Red 52 is a fluorescent rhodamine dye also known as Sulforhodamine B and Kiton Red, and used as a polar tracer.

Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Synonyms:  Food Red No.106, Sodium 4-(3,6-bis(diethylamino)xanthylium-9-yl)benzene-1,3-disulphonate, Kiton Red S, Sulphorhodamine B sodium salt, Lissamine rhodamine B-200
  • CAS Number:3520-42-1
  • Formula:C₂₇H₃₀N₂O₇S₂
  • MDL Number:MFCD00010180
  • Molecular Weight:580.65 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pk:
    5 kg

Specifications

Consigli

I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".

Altrimenti riceverai suggerimenti generici..