Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Dexamethasone
Dexamethasone ≥97%, micronised powder USP
Dexamethasone ≥97%, micronised powder USP
SPCMDE121-5GM
:
Spectrum Chemical
:
Dexamethasone ≥97%, micronised powder USP
SPCMDE121-5GM
:
Spectrum Chemical
:
Pack type:
Glass, amber
Pk:
5 g
Seq:
3173
Assay:
97% Min.
:
:
392.47 g/mol
:
268…271 °C
:
Ambient
:
200-003-9
:
50-02-2
:
C₂₂H₂₉FO₅
:
MFCD00064136