Sodium acetate, anhydrous ≥99.0% USP
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:127-09-3
:
Formula:H₃CCOONa
MDL Number:MFCD00012459
:82.03 g/mol
Storage Temperature:Ambient
Specification Test Results
- Assay (Dried Basis):99.0 - 101.0%
- pH of a 3% Solution @ 25°C:7.5 - 9.2
- Loss On Drying:max. 1.0 %
- Insoluble Matter:max. 0.05 %
- Chloride (Cl):max. 350 ppm
Specification Test Results
Frequently Bought Together