- Antibody type:Primary
- Antigen name:C3orf20
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:601-700/904
- Form:Liquid
- Antigen synonyms:uncharacterized protein C3orf20 isoform 1|C3orf20|Chromosome 3 open reading frame 20|Uncharacterized protein C3orf20|CC020_HUMAN|DKFZP434N1817
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
- Molecular weight:101 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human C3orf20
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
C3orf20 (chromosome 3 open reading frame 20), also known as DKFZp434N1817, is a 904 amino acid single-pass membrane protein that exists as two alternatively spliced isoforms. C3orf20 is encoded by a gene that maps to human chromosome 3p25.1. Chromosome 3 is made up of approximately 214 million bases encoding over 1,100 genes. Notably, there is a chemokine receptor gene cluster and a variety of human cancer related loci on chromosome 3. Particular regions of the chromosome 3 short arm are deleted in many types of cancer cells. Key tumor suppressing genes on chromosome 3 encode apoptosis mediator RASSF1, cell migration regulator HYAL1 and angiogenesis suppressor SEMA3B. Marfan Syndrome, porphyria, von Hippel-Lindau syndrome, osteogenesis imperfecta and Charcot-Marie-Tooth disease are a few of the numerous genetic diseases associated with chromosome 3.
Type: Primary
Antigen: C3orf20
Clonality: Polyclonal
Clone:
Conjugation: HRP
Public Immunogen Range: 601-700/904
Host: Rabbit
Isotype: IgG
Reactivity: