Order Entry
Italy
ContactUsLinkComponent
Anti-C20orf152 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anti-C20orf152 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-15097R-A647
Supplier:  Bioss
Anti-C20orf152 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-15097R-A647
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    C20orf152
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 647
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    21-120/576
  • Form:
    Liquid
  • Gene ID:
    140894
  • Antigen synonyms:
    Uncharacterized protein C20orf152 homolog|Uncharacterized protein C20orf152|Chromosome 20 open reading frame 152|RP5-954P9.1|C20orf152|CT152_HUMAN|dJ954P9.1
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
  • Molecular weight:
    68 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human C20orf152
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

Representing about 2% of human DNA, chromosome 20 consists of approximately 63 million bases and 600 genes. Chromosome 20 contains a region with numerous genes expressed in the epididymis, which are thought important for seminal production, and some viewed as potential targets for male contraception. The PRNP gene encoding the prion protein associated with spongiform encephalopathies, like Creutzfeldt-Jakob disease, is found on chromosome 20. Amyotrophic lateral sclerosis, spinal muscular atrophy, ring chromosome 20 epilepsy syndrome and Alagille syndrome are also associated with chromosome 20. The C20orf152 gene product has been provisionally designated C20orf152 pending further characterization.

Type: Primary
Antigen: C20orf152
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 647
Public Immunogen Range: 21-120/576
Host: Rabbit
Isotype: IgG
Reactivity: Human