Anti-CHPF Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-13928R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:CHPF
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:51-150/775
- Form:Liquid
- Gene ID:79586
- Antigen synonyms:Chondroitin sulfate synthase 2, Chondroitin-polymerizing factor, Glucuronosyl-N-acetylgalactosaminyl-proteoglycan 4-beta-N-acetylgalactosaminyltransferase II,
N-acetylgalactosaminyl-proteoglycan 3-beta-glucuronosyltransferase II,
N-acetylgalactosaminyltransferase 2 - Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
- Molecular weight:85 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human CHPF
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Has both beta-1,3-glucuronic acid and beta-1,4-N-acetylgalactosamine transferase activity. Transfers glucuronic acid (GlcUA) from UDP-GlcUA and N-acetylgalactosamine (GalNAc) from UDP-GalNAc to the non-reducing end of the elongating chondroitin polymer. Isoform 2 may facilitate PARK2 transport into the mitochondria. In collaboration with PARK2, isoform 2 may enhance cell viability and protect cells from oxidative stress.
Type: Primary
Antigen: CHPF
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 51-150/775
Host: Rabbit
Isotype: IgG
Reactivity: Human