Order Entry
Italy
ContactUsLinkComponent
Anti-Claudin 16 Rabbit Polyclonal Antibody (Cy5®)
Anti-Claudin 16 Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-13752R-CY5
Supplier:  Bioss
Anti-Claudin 16 Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-13752R-CY5
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Claudin 16
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy5®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Epitope:
    95-150/305
  • Form:
    Liquid
  • Gene ID:
    10686
  • Antigen synonyms:
    HOMG3|PCLN-1|Paracellin-1|Claudin-16|CLDN16|PCLN1
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
  • Molecular weight:
    34 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human Claudin 16
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

Plays a major role in tight junction-specific obliteration of the intercellular space, through calcium-independent cell-adhesion activity. Involved in paracellular magnesium reabsorption. Required for a selective paracellular conductance. May form, alone or in partnership with other constituents, an intercellular pore permitting paracellular passage of magnesium and calcium ions down their electrochemical gradients. Alternatively, it could be a sensor of magnesium concentration that could alter paracellular permeability mediated by other factors.

Type: Primary
Antigen: Claudin 16
Clonality: Polyclonal
Clone:
Conjugation: Cy5
Public Immunogen Range: 95-150/305
Host: Rabbit
Isotype: IgG
Reactivity: