Specifications
- Antibody type:Primary
- Antigen name:Protocadherin 10
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat
- Western blot:Yes
- Epitope:21-120/1040
- Form:Liquid
- Gene ID:57575
- Antigen synonyms:KIAA14|PCDH19|OL-PCDH|PCDH1|Protocadherin-1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
- Molecular weight:110 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human PCDH10
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Protocadherins are a large family of cadherin-like cell adhesion proteins that are involved in the establishment and maintenance of neuronal connections in the brain. There are three protocadherin gene clusters, designated alpha, beta and gamma, all of which contain multiple tandemly arranged genes. PCDH10 (protocadherin 10), also known as PCDH19 or OL-PCDH, is a 1,040 amino acid single-pass type I membrane protein that contains six cadherin domains and one transmembrane domain. Expressed at moderate levels in brain, ovary and testis and present at lower levels in all other tissues, PCDH10 functions as a calcium-dependent cell-adhesion protein that may function as a tumor suppressor. When underexpressed, PCDH10 is associated with the progression of various carcinomas, including gastric cancer.
Type: Primary
Antigen: Protocadherin 10
Clonality: Polyclonal
Clone:
Conjugation: Cy5
Public Immunogen Range: 21-120/1040
Host: Rabbit
Isotype: IgG
Reactivity: Rat