Order Entry
Italy
ContactUsLinkComponent
Anti-FBXO38 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anti-FBXO38 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-9122R-A750
Supplier:  Bioss
Anti-FBXO38 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-9122R-A750
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    FBXO38
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 750
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    81545
  • Antigen synonyms:
    MoKA|Fbx38|FBXO 38|Modulator of KLF7 activity homolog|F box only protein 38|FBX38_HUMAN|FBXO38|F box protein 38
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Storage temperature:
    Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human FBXO38
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

F box protein 38 probably recognises and binds to some phosphorylated proteins and promotes their ubiquitination and degradation. It may coactivate KLF7, but does not seem to promote KLF7 ubiquitination. There are three named isoforms.

Type: Primary
Antigen: FBXO38
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat