Anti-DBF4A Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-7895R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:DBF4A
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:21-110/674
- Form:Liquid
- Gene ID:10926
- Antigen synonyms:Activator of S phase kinase|DBF4A|Protein DBF4 homolog A|DBF4A_HUMAN|DBF4|DBF4 type zinc finger containing protein 1|DBF4-type zinc finger-containing protein 1|ASK|ZDBF1|Chfon homolog A
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:77 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human DBF4A
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Regulatory subunit for CDC7 which activates its kinase activity thereby playing a central role in DNA replication and cell proliferation. Required for progression of S phase. The complex CDC7-DBF4A selectively phosphorylates MCM2 subunit at 'Ser-40' and 'Ser-53' and then is involved in regulating the initiation of DNA replication during cell cycle.Tissue specificity: Highly expressed in testis and thymus. Expressed also in most cancer cells lines.
Type: Primary
Antigen: DBF4A
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 21-110/674
Host: Rabbit
Isotype: IgG
Reactivity: