Order Entry
Italy
ContactUsLinkComponent
 
Anti-CstF-64 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-6995R-A680
 :  Bioss
Anti-CstF-64 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-6995R-A680
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    CstF-64
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Epitope:
    61-160/616
  • Form:
    Liquid
  • Gene ID:
    23283
  • Antigen synonyms:
    CF1 64 kDa subunit, tau variant|Cleavage stimulation factor 64 kDa subunit, tau variant|TauCstF64|KIAA0689|CSTF 64 kDa subunit, tau variant|CSTFT_HUMAN
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    64 kDa
  • Storage temperature:
    Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human CSTF2T/CstF-64
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

 

 

Polyadenylation of mRNA precursors is a two-step reaction that requires multiple protein factors. The first step, endonucleolytic cleavage of polyadenylation substrates, requires CstF (cleavage stimulation factor), a heterotrimer that is composed of three distinct subunits. CstF-64 contains an RNA binding domain and is responsible for the RNA binding activity of CstF. CstF-64 is expressed in all somatic cells and in pre- and postmeiotic, but not meiotic, germ cells. However, a large variant of CstF-64, called t CstF-64, is abundantly expressed in meiotic and postmeiotic cells in the testis and to a lesser extent in the brain, and promotes the germ cell pattern of polyadenylation. The gene encoding CstF-64 (designated CSTF2) maps to the X chromosome, whereas t CstF-64 is encoded by an autosomal gene. The increase in CstF-64 concentration during B cell activation switches IgM heavy chain mRNA expression from membrane-bound to secreted forms, suggesting that CstF-64 plays a key role in regulating IgM heavy chain expression during B cell differentiation.

Type: Primary
Antigen: CstF-64
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 61-160/616
Host: Rabbit
Isotype: IgG
Reactivity: