Anti-BCA2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
BOSSBS-6757R-A680
: Bioss
- Antibody type:Primary
- Antigen name:BCA2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:161-260/304
- Form:Liquid
- Antigen synonyms:Rabring 7|Zinc finger protein 364|Breast cancer associated gene 2|RING finger protein 115|RN115_HUMAN|RNF115|BCA2|CL469780
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:34 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human BCA2
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
The RING finger family of proteins possess ubiquitin ligase activity and play pivotal roles in protein degradation and receptor-mediated endocytosis. The autoubiquitination activity of ZNF364 indicates that it is a novel RING-type E3 ligase. Its effects on cell growth indicate that ZNF364 may be important for the ubiquitin modification of proteins crucial to breast carcinogenesis and growth.
Type: Primary
Antigen: BCA2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 161-260/304
Host: Rabbit
Isotype: IgG
Reactivity: