Anti-DEPDC1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
BOSSBS-6525R-A680
: Bioss
- Antibody type:Primary
- Antigen name:DEPDC1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Epitope:606-660/811
- Form:Liquid
- Gene ID:55635
- Antigen synonyms:Cell cycle control protein SDP35|DEP domain containing protein 1A|DEPDC 1A|DEP domain containing 1|DEP.8|DEPDC 1|DEPDC1-V2|DEP domain-containing protein 1A|SDP35|SDP 35|FLJ20354|Depdc1a|DEP1A_HUMAN|DEPDC1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:93 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human DEPDC1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
DEPDC1.
Type: Primary
Antigen: DEPDC1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 606-660/811
Host: Rabbit
Isotype: IgG
Reactivity: Mouse