Anti-CGR19 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-6373R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:CGR19
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Epitope:45-150/332
- Form:Liquid
- Gene ID:10668
- Antigen synonyms:Cell growth regulatory with ring finger domain|CGRRF-1|RING finger protein 197|CGRRF1|CGRRF 1|Cell growth regulator with RING finger domain protein 1|Cell growth regulatory gene 19 protein|RNF197|CGRF1_HUMAN|Cell growth regulator with ring finger domain 1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:38 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human CGR19
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
CGR19 is involved in the negative regulation of cell proliferation and cell cycle arrest in response to stress.
Type: Primary
Antigen: CGR19
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 45-150/332
Host: Rabbit
Isotype: IgG
Reactivity: Mouse