Anti-Glutamine PRPP amidotransferase Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-6359R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Glutamine PRPP amidotransferase
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Epitope:351-430/517
- Form:Liquid
- Gene ID:5471
- Antigen synonyms:Amidophosphoribosyltransferase|PPAT|Glutamine phosphoribosylpyrophosphate amidotransferase|GPAT|ATASE|PRAT
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:56 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Glutamine PRPP amidotransferase
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Phosphoribosyl pyrophosphate amidotransferase (PPAT) is a regulatory allosteric enzyme that catalyzes the first step of de novo purine nucleotide biosynthesis. The genes for PPAT and PAICS/AIRC, (a bifunctional enzyme catalyzing steps six and seven in the purine nucleotide biosynthesis pathway), are located in close proximity on chromosome 4.
Type: Primary
Antigen: Glutamine PRPP amidotransferase
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 351-430/517
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat